Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bcor
Entrez Gene ID
71458
Gene Name
BCL6 interacting corepressor
Gene Aliases
5830466J11Rik, 8430401K06Rik, BcorR, D930024N20Rik, mKIAA1575
Chromosome Location
Chr.X: 12036737 - 12160355 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168321.1 NP_001161793.1 2 - 3 192 55
NM_029510.3 NP_083786.2 2 - 3 861 55
NM_175044.3 NP_778209.2 2 - 3 861 55
NM_175045.3 NP_778210.2 2 - 3 861 55
NM_175046.3 NP_778211.2 2 - 3 861 55
GenBank mRNA AK053309.1 - 2 - 3 862 55
AK086385.2 - 2 - 3 368 55
AK129398.1 - 2 - 3 192 55
AY161170.1 - 2 - 3 176 55
AY161171.1 - 2 - 3 176 55
AY161172.1 - 2 - 3 176 55
AY161173.1 - 2 - 3 176 55
BC058656.1 - 2 - 3 362 55

View Full Product Details