Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bcor
Entrez Gene ID
71458
Gene Name
BCL6 interacting corepressor
Gene Aliases
5830466J11Rik, 8430401K06Rik, BcorR, D930024N20Rik, mKIAA1575
Chromosome Location
Chr.X: 12036737 - 12160355 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168321.1 NP_001161793.1 9 - 10 4390 112
NM_029510.3 NP_083786.2 10 - 11 5215 112
NM_175044.3 NP_778209.2 9 - 10 5161 112
NM_175045.3 NP_778210.2 10 - 11 5113 112
NM_175046.3 NP_778211.2 9 - 10 5059 112
GenBank mRNA AK018370.1 - 2 - 3 470 112
AK031282.1 - 2 - 3 518 112
AK129398.1 - 9 - 10 4390 112
AY161170.1 - 10 - 11 4530 112
AY161171.1 - 9 - 10 4476 112
AY161172.1 - 10 - 11 4428 112
AY161173.1 - 9 - 10 4374 112
BC038637.1 - 2 - 3 420 112
BC058656.1 - 10 - 11 4614 112

View Full Product Details