Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gm31063
Entrez Gene ID
102633167
Gene Name
predicted gene, 31063
Gene Aliases
-
Chromosome Location
Chr.12: 65190613 - 65213336 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK081945.1 - - 175 68


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB377327.2 - 1 - 2 213 Chr. 12 - 65187872 - 65213501 [-] on Build GRCm38 68
GenBankmRNA BB649741.1 - 1 - 2 175 Chr. 12 - 65187867 - 65213501 [-] on Build GRCm38 68


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream MIS18 binding protein 1 Mis18bp1 6720403H11, C79407, Knl2, mKIAA1903 -29.9kb Sense Chr. - 65132734 - 65172580 on Build GRCm38
Downstream WD repeat domain 20, retrogene Wdr20rt 4921538B03Rik, 4930427E19Rik, Wdr20, Wdr20b 12.0kb Anti-sense Chr. - 65225517 - 65228454 on Build GRCm38

View Full Product Details