Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Acmsd
Entrez Gene ID
266645
Gene Name
amino carboxymuconate semialdehyde decarboxylase
Gene Aliases
-
Chromosome Location
Chr.1: 127729413 - 127767564 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033041.2 NP_001028213.1 3 - 4 274 87
GenBank mRNA AB071419.1 - 3 - 4 201 87
AK143962.1 - 3 - 4 276 87
BC139281.1 - 3 - 4 225 87

View Full Product Details