The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011783.2 | NP_035913.1 | 6 - 7 | 444 | 92 |
GenBank mRNA | AB016592.1 | - | 6 - 7 | 443 | 92 |
AF044262.1 | - | 6 - 7 | 444 | 92 | |
AK007677.1 | - | 6 - 7 | 463 | 92 | |
AK008296.1 | - | 5 - 6 | 512 | 92 | |
BC013334.1 | - | 6 - 7 | 462 | 92 |