Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Mfsd4
- Entrez Gene ID
-
213006
- Gene Name
- major facilitator superfamily domain containing 4
- Gene Aliases
-
A230072B04, A930031D07Rik, AI850289
- Chromosome Location
-
Chr.1: 132022806 - 132068062 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK048692.1 |
- |
1 - 2 |
400 |
104 |
View Full Product Details