The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028749.1 | NP_083025.1 | 11 - 12 | 856 | 79 |
GenBank mRNA | AK002734.1 | - | 11 - 12 | 856 | 79 |
BC022734.1 | - | 11 - 12 | 852 | 79 |