Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Plekhh3
Entrez Gene ID
217198
Gene Name
pleckstrin homology domain containing, family H (with MyTH4 domain) member 3
Gene Aliases
BC020025
Chromosome Location
Chr.11: 101162679 - 101171405 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146030.2 NP_666142.1 11 - 12 2427 74
GenBank mRNA AK164128.1 - 11 - 12 2437 74

View Full Product Details