Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mlx
Entrez Gene ID
21428
Gene Name
MAX-like protein X
Gene Aliases
Tcfl4, bHLHd13
Chromosome Location
Chr.11: 101086709 - 101092456 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159385.1 NP_001152857.1 3 - 4 201 63
NM_011550.3 NP_035680.3 3 - 4 363 63
GenBank mRNA AK142605.1 - 3 - 4 167 63
BC038004.1 - 3 - 4 343 63
BC080768.1 - 2 - 3 1168 63
CK620266.1 - 3 - 4 205 63

View Full Product Details