The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001165253.1 | NP_001158725.1 | 1 - 2 | 154 | 61 |
GenBank mRNA | AK157358.1 | - | 1 - 2 | 155 | 61 |
BQ571563.1 | - | 1 - 2 | 153 | 61 |