The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001165253.1 | NP_001158725.1 | 5 - 6 | 509 | 106 |
NM_001165254.1 | NP_001158726.1 | 5 - 6 | 508 | 106 | |
NM_146034.3 | NP_666146.3 | 7 - 8 | 607 | 106 | |
GenBank mRNA | AK029466.1 | - | 5 - 6 | 508 | 106 |
AK053010.1 | - | 6 - 7 | 514 | 106 | |
AK080312.1 | - | 6 - 7 | 556 | 106 | |
AK087188.1 | - | 6 - 7 | 522 | 106 | |
AK157358.1 | - | 5 - 6 | 510 | 106 | |
BC026864.1 | - | 7 - 8 | 559 | 106 | |
BQ571563.1 | - | 5 - 6 | 508 | 106 |