The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_120500.1 | 14 - 15 | 2314 | 72 | |
GenBank mRNA | AK006249.1 | - | 12 - 13 | 1580 | 72 |
AY461596.1 | - | 14 - 15 | 2332 | 72 |