Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mael
Entrez Gene ID
98558
Gene Name
maelstrom homolog (Drosophila)
Gene Aliases
4933405K18Rik, AU019877
Chromosome Location
Chr.1: 166201385 - 166238744 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175296.4 NP_780505.1 6 - 7 726 83
GenBank mRNA AK077103.1 - 6 - 7 723 83
AK165801.1 - 6 - 7 723 83
BC050800.1 - 6 - 7 726 83

View Full Product Details