Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Id2
Entrez Gene ID
15902
Gene Name
inhibitor of DNA binding 2
Gene Aliases
AI255428, C78922, Idb2, bHLHb26
Chromosome Location
Chr.12: 25093799 - 25096092 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010496.3 NP_034626.1 1 - 2 433 84
GenBank mRNA AK003222.1 - 1 - 2 430 84
AK013378.1 - 1 - 2 430 84
AK135547.1 - 1 - 2 435 84
AK146346.1 - 1 - 2 433 84
AK146606.1 - 1 - 2 436 84
BC006921.1 - 1 - 2 410 84
BC053699.1 - 1 - 2 420 84
M69293.1 - 1 - 2 427 84

View Full Product Details