The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027941.1 | NP_082217.1 | 1 - 2 | 149 | 56 |
GenBank mRNA | AK005720.1 | - | 1 - 2 | 201 | 56 |
BC100519.1 | - | 1 - 2 | 149 | 56 | |
JF934738.1 | - | 1 - 2 | 100 | 56 |