Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrrc31
Entrez Gene ID
320352
Gene Name
leucine rich repeat containing 31
Gene Aliases
E230002P03Rik
Chromosome Location
Chr.3: 30678917 - 30699840 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK136409.1 - 3 - 4 408 75

View Full Product Details