Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tagln2
Entrez Gene ID
21346
Gene Name
transgelin 2
Gene Aliases
2700094C18Rik, SM22beta, Sm22B, Sm22a
Chromosome Location
Chr.1: 172500246 - 172507375 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178598.2 NP_848713.1 4 - 5 573 87
GenBank mRNA AF149291.1 - 4 - 5 564 87
AF465519.1 - 4 - 5 510 87
AK151576.1 - 4 - 5 573 87
AK167395.1 - 4 - 5 558 87
AK167922.1 - 4 - 5 555 87
AK171088.1 - 4 - 5 541 87
AK172892.1 - 4 - 5 512 87
BC009076.1 - 4 - 5 555 87
BC049861.1 - 4 - 5 772 87

View Full Product Details