Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slamf6
Entrez Gene ID
30925
Gene Name
SLAM family member 6
Gene Aliases
KAL1, KAL1b, Ly108, NTB-A, NTBA, SF2000
Chromosome Location
Chr.1: 171917199 - 171953170 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030710.2 NP_109635.1 6 - 7 1083 119

View Full Product Details