Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slamf8
Entrez Gene ID
74748
Gene Name
SLAM family member 8
Gene Aliases
5830408F06Rik, Blame, SBBI42
Chromosome Location
Chr.1: 172581377 - 172591191 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AB196828.1 - 2 - 3 357 73
AB196829.1 - 2 - 3 357 73
AB196830.1 - 2 - 3 357 73
AB196831.1 - 2 - 3 357 73
AK156077.1 - 2 - 3 392 73

View Full Product Details