Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slamf8
Entrez Gene ID
74748
Gene Name
SLAM family member 8
Gene Aliases
5830408F06Rik, Blame, SBBI42
Chromosome Location
Chr.1: 172581377 - 172591191 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029084.3 NP_083360.2 3 - 4 808 66
GenBank mRNA AB196828.1 - 3 - 4 672 66
AB196829.1 - 3 - 4 672 66
AB196830.1 - 3 - 4 672 66
AB196831.1 - 3 - 4 672 66
AK017911.1 - 3 - 4 711 66
AK142633.1 - 3 - 4 809 66
AK156077.1 - 3 - 4 707 66

View Full Product Details