Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cyp7b1
Entrez Gene ID
13123
Gene Name
cytochrome P450, family 7, subfamily b, polypeptide 1
Gene Aliases
AW261589, D3Ertd552e, hct-1
Chromosome Location
Chr.3: 18071950 - 18243893 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007825.4 NP_031851.3 3 - 4 981 90
GenBank mRNA AK013034.1 - 3 - 4 911 90
AK140100.1 - 3 - 4 973 90
AK172152.1 - 3 - 4 982 90
BC038810.1 - 3 - 4 941 90

View Full Product Details