Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Optc
Entrez Gene ID
269120
Gene Name
opticin
Gene Aliases
-
Chromosome Location
Chr.1: 133897193 - 133909714 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160420.2 NP_001153892.1 3 - 4 415 91
NM_001160421.2 NP_001153893.1 2 - 3 526 91
NM_001160422.1 NP_001153894.1 2 - 3 526 91
NM_054076.3 NP_473417.2 2 - 3 526 91
GenBank mRNA AF333980.1 - 2 - 3 525 91
AK053318.1 - 2 - 3 309 91
AK153750.1 - 3 - 4 416 91
AY077682.1 - 2 - 3 518 91
BC099435.1 - 2 - 3 359 91
BC115928.1 - 2 - 3 479 91

View Full Product Details