The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284525.1 | NP_001271454.1 | 11 - 12 | 1417 | 69 |
NM_027979.2 | NP_082255.1 | 11 - 12 | 1381 | 69 | |
GenBank mRNA | AY458654.1 | - | 11 - 12 | 1381 | 69 |
AY536287.1 | - | 10 - 11 | 1255 | 69 | |
BC138765.1 | - | 11 - 12 | 1383 | 69 |