Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gmps
Entrez Gene ID
229363
Gene Name
guanine monophosphate synthetase
Gene Aliases
AA591640, AI047208
Chromosome Location
Chr.3: 63964819 - 64021490 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033300.2 NP_001028472.2 11 - 12 1578 147
GenBank mRNA AK049755.1 - 11 - 12 1542 147
AK143197.1 - 11 - 12 1579 147
AK146654.1 - 11 - 12 1596 147
AK167857.1 - 11 - 12 1555 147
AK168239.1 - 11 - 12 1528 147
AK169043.1 - 11 - 12 1562 147
AK169701.1 - 11 - 12 1576 147
BC037232.1 - 9 - 10 1230 147
BC080685.1 - 6 - 7 790 147
BC138411.1 - 11 - 12 1534 147
BC138412.1 - 11 - 12 1534 147

View Full Product Details