The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_206534.1 | NP_996257.1 | 2 - 3 | 205 | 80 |
GenBank mRNA | AK136890.1 | - | 1 - 2 | 4909 | 80 |
BC076499.1 | - | 2 - 3 | 205 | 80 |