Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pxdn
Entrez Gene ID
69675
Gene Name
peroxidasin homolog (Drosophila)
Gene Aliases
2310075M15Rik, C85409, E330004E07, VPO1, mKIAA0230
Chromosome Location
Chr.12: 29937877 - 30017658 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181395.2 NP_852060.2 7 - 8 821 61
GenBank mRNA AK054255.1 - 6 - 7 736 61
AK122223.1 - 7 - 8 591 61
AK142872.1 - 7 - 8 821 61
AK163597.1 - 7 - 8 808 61
BC150789.1 - 7 - 8 816 61

View Full Product Details