Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pvrl4
Entrez Gene ID
71740
Gene Name
poliovirus receptor-related 4
Gene Aliases
1200017F15Rik, Prr4
Chromosome Location
Chr.1: 171370173 - 171388287 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122680.1 NP_001116152.1 4 - 5 1344 101
NM_027893.3 NP_082169.2 4 - 5 1344 101
GenBank mRNA AF472510.1 - 4 - 5 849 101
AK004821.1 - 4 - 5 1163 101
AK154983.1 - 5 - 6 1042 101
BC024948.1 - 4 - 5 1100 101

View Full Product Details