Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pvrl4
Entrez Gene ID
71740
Gene Name
poliovirus receptor-related 4
Gene Aliases
1200017F15Rik, Prr4
Chromosome Location
Chr.1: 171370173 - 171388287 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027893.3 NP_082169.2 7 - 8 1723 69
GenBank mRNA AF472510.1 - 7 - 8 1228 69
AK028479.1 - 7 - 8 1499 69
AK154983.1 - 8 - 9 1421 69
BC024948.1 - 7 - 8 1479 69

View Full Product Details