Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Atf6
Entrez Gene ID
226641
Gene Name
activating transcription factor 6
Gene Aliases
9130025P16Rik, 9630036G24, AA789574, Atf6alpha, ESTM49
Chromosome Location
Chr.1: 170704457 - 170867771 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081304.1 NP_001074773.1 15 - 16 1837 114
GenBank mRNA AK036106.1 - 15 - 16 1816 114
AK036335.1 - 15 - 16 1835 114
AK087000.1 - 15 - 16 1823 114
BC043662.1 - 14 - 15 1684 114
BC141028.1 - 15 - 16 1831 114

View Full Product Details