The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178767.5 | NP_848882.2 | 12 - 13 | 1661 | 87 |
GenBank mRNA | AK040702.1 | - | 12 - 13 | 1662 | 87 |