The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008510.1 | NP_032536.1 | 2 - 3 | 189 | 109 |
GenBank mRNA | AK079820.1 | - | 2 - 3 | 202 | 109 |
BC062249.1 | - | 2 - 3 | 189 | 109 | |
D43769.1 | - | 2 - 3 | 189 | 109 | |
U15607.1 | - | 2 - 3 | 187 | 109 |