Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fcer1a
Entrez Gene ID
14125
Gene Name
Fc receptor, IgE, high affinity I, alpha polypeptide
Gene Aliases
FcERI, Fce1a, Fcr-5, fcepsilonri
Chromosome Location
Chr.1: 173221269 - 173227232 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010184.2 NP_034314.2 4 - 5 641 120
GenBank mRNA BC125455.1 - 4 - 5 606 120
BC125457.1 - 4 - 5 605 120
BC145138.1 - 4 - 5 563 120
J05018.1 - 4 - 5 626 120

View Full Product Details