The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010941.3 | NP_035071.3 | 1 - 2 | 176 | 112 |
GenBank mRNA | AF100198.1 | - | 1 - 2 | 176 | 112 |
AK013945.1 | - | 1 - 2 | 59 | 112 | |
AK088026.1 | - | 1 - 2 | 59 | 112 | |
AK140930.1 | - | 1 - 2 | 57 | 112 | |
BC019945.1 | - | 1 - 2 | 45 | 112 | |
BC052834.1 | - | 1 - 2 | 137 | 112 |