Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cacybp
Entrez Gene ID
12301
Gene Name
calcyclin binding protein
Gene Aliases
SIP
Chromosome Location
Chr.1: 160202367 - 160212892 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009786.2 NP_033916.1 1 - 2 251 125
GenBank mRNA AK013924.1 - 1 - 2 182 125
AK089043.1 - 1 - 2 105 125
AK145013.1 - 1 - 2 218 125
BC025948.1 - 1 - 2 136 125
U97327.1 - 1 - 2 81 125

View Full Product Details