Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zbtb41
Entrez Gene ID
226470
Gene Name
zinc finger and BTB domain containing 41 homolog
Gene Aliases
8430415N23Rik, 9430031N01, 9830132G07Rik, AI316857
Chromosome Location
Chr.1: 139421719 - 139453007 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172643.5 NP_766231.2 6 - 7 1824 97
GenBank mRNA AK034756.1 - 5 - 6 921 97
AK036544.1 - 6 - 7 1823 97
AK079719.1 - 2 - 3 182 97
AK163302.1 - 5 - 6 1097 97
BC046465.1 - 5 - 6 993 97
BC150859.1 - 5 - 6 1163 97
BC172012.1 - 6 - 7 1089 97

View Full Product Details