Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Krt4
Entrez Gene ID
16682
Gene Name
keratin 4
Gene Aliases
AW108092, K4, Krt-2.4, Krt2-4
Chromosome Location
Chr.15: 101918535 - 101924735 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008475.2 NP_032501.2 8 - 9 1476 73
GenBank mRNA BC064008.1 - 8 - 9 1457 73
BC145839.1 - 8 - 9 1431 73
BC145841.1 - 8 - 9 1431 73

View Full Product Details