The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010517.3 | NP_034647.1 | 1 - 2 | 587 | 61 |
GenBank mRNA | AK031212.1 | - | 1 - 2 | 604 | 61 |
AK150229.1 | - | 1 - 2 | 600 | 61 | |
AK165688.1 | - | - | 605 | 61 | |
BC019836.1 | - | 1 - 2 | 587 | 61 | |
X81582.1 | - | 1 - 2 | 536 | 61 |