The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080925.1 | NP_001074394.1 | 4 - 5 | 1195 | 70 |
GenBank mRNA | BC080279.1 | - | 3 - 4 | 263 | 70 |
BC130266.1 | - | 5 - 6 | 471 | 70 |