The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159748.1 | NP_001153220.1 | 1 - 2 | 276 | 78 |
NM_011459.4 | NP_035589.1 | 1 - 2 | 276 | 78 | |
GenBank mRNA | AK032659.1 | - | 1 - 2 | 277 | 78 |
AK036811.1 | - | 1 - 2 | 212 | 78 | |
AK048230.1 | - | 1 - 2 | 236 | 78 | |
AK148413.1 | - | 1 - 2 | 274 | 78 | |
BC025121.1 | - | 1 - 2 | 266 | 78 | |
U96703.2 | - | 1 - 2 | 267 | 78 |