The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026313.1 | NP_080589.1 | 10 - 11 | 1401 | 67 |
GenBank mRNA | AK151228.1 | - | 10 - 11 | 1449 | 67 |
AK151839.1 | - | 10 - 11 | 1449 | 67 | |
BC009092.1 | - | 10 - 11 | 1401 | 67 |