Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fmnl3
Entrez Gene ID
22379
Gene Name
formin-like 3
Gene Aliases
2700073B04Rik, FBP11, Wbp3, mKIAA2014
Chromosome Location
Chr.15: 99317210 - 99370503 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011711.1 NP_035841.1 11 - 12 1242 96
GenBank mRNA AK129473.1 - 11 - 12 1161 96
AK139711.1 - 11 - 12 1225 96
AK143308.1 - 11 - 12 1141 96
AK155331.1 - 11 - 12 1259 96
AK163361.1 - 11 - 12 1228 96
AK170744.1 - 10 - 11 1072 96
BC023797.1 - 10 - 11 962 96
BC085094.1 - 11 - 12 1139 96
BC131961.1 - 11 - 12 1118 96

View Full Product Details