The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011711.1 | NP_035841.1 | 14 - 15 | 1770 | 142 |
GenBank mRNA | AK129473.1 | - | 14 - 15 | 1689 | 142 |
AK139711.1 | - | 14 - 15 | 1754 | 142 | |
AK143308.1 | - | 14 - 15 | 1670 | 142 | |
AK155331.1 | - | 14 - 15 | 1787 | 142 | |
AK170744.1 | - | 13 - 14 | 1600 | 142 | |
BC023797.1 | - | 13 - 14 | 1490 | 142 | |
BC085094.1 | - | 14 - 15 | 1665 | 142 | |
BC131961.1 | - | 14 - 15 | 1646 | 142 | |
U92453.1 | - | 3 - 4 | 444 | 141 |