Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Osbpl7
Entrez Gene ID
71240
Gene Name
oxysterol binding protein-like 7
Gene Aliases
4933437E18Rik
Chromosome Location
Chr.11: 97050344 - 97068904 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081434.1 NP_001074903.1 10 - 11 1088 88
GenBank mRNA AK038088.1 - 5 - 6 1675 88
AK171585.1 - 10 - 11 1114 88
AK219892.1 - 3 - 4 298 88

View Full Product Details