Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prkaa1
Entrez Gene ID
105787
Gene Name
protein kinase, AMP-activated, alpha 1 catalytic subunit
Gene Aliases
AI194361, AI450832, AL024255, AMPKalpha1, C130083N04Rik
Chromosome Location
Chr.15: 5143861 - 5181899 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013367.3 NP_001013385.3 3 - 4 375 121
GenBank mRNA AK147811.1 - 3 - 4 352 121
AK160612.1 - 3 - 4 356 121
AY885266.1 - 3 - 4 334 121
BC086695.1 - 3 - 4 345 121

View Full Product Details