Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Adamts20
Entrez Gene ID
223838
Gene Name
a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 20
Gene Aliases
Adamst20, bt
Chromosome Location
Chr.15: 94268314 - 94465427 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164785.1 NP_001158257.1 3 - 4 869 64
NM_001164786.1 NP_001158258.1 3 - 4 869 64
NM_177431.4 NP_803180.3 3 - 4 869 64
GenBank mRNA AJ512753.3 - 3 - 4 638 64
AK145288.1 - 3 - 4 870 64
AY189815.1 - 3 - 4 777 64
AY189816.1 - 3 - 4 777 64
BC121792.1 - 3 - 4 708 64

View Full Product Details