Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nxf3
Entrez Gene ID
245610
Gene Name
nuclear RNA export factor 3
Gene Aliases
Gm384
Chromosome Location
Chr.X: 136072099 - 136085553 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024141.4 NP_001019312.1 13 - 14 1296 127
GenBank mRNA AB178899.1 - 13 - 14 1299 127

View Full Product Details