Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C7
Entrez Gene ID
109828
Gene Name
complement component 7
Gene Aliases
A530026G17, Gm1238
Chromosome Location
Chr.15: 4988762 - 5063773 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243837.1 NP_001230766.1 4 - 5 398 92

View Full Product Details