The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008002.4 | NP_032028.1 | 2 - 3 | 1121 | 145 |
GenBank mRNA | AK034742.1 | - | 2 - 3 | 687 | 145 |
AK045267.1 | - | 2 - 3 | 1122 | 145 | |
AK085244.1 | - | 2 - 3 | 443 | 145 | |
AK141633.1 | - | 2 - 3 | 443 | 145 | |
BC048229.1 | - | 2 - 3 | 786 | 145 | |
D89080.1 | - | 2 - 3 | 436 | 145 | |
U94517.1 | - | 2 - 3 | 436 | 145 |