Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gramd4
Entrez Gene ID
223752
Gene Name
GRAM domain containing 4
Gene Aliases
9930016O13
Chromosome Location
Chr.15: 86057695 - 86137636 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205353.1 NP_001192282.1 5 - 6 765 86
NM_172611.4 NP_766199.1 5 - 6 667 86
GenBank mRNA AK036832.1 - 5 - 6 656 86
AK129209.1 - 5 - 6 569 86
AK142266.1 - 5 - 6 620 86
BC021523.1 - 1 - 2 41 86
BC085148.1 - 5 - 6 723 86
BC139228.1 - 5 - 6 656 86
BC139229.1 - 5 - 6 656 86

View Full Product Details