The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_207267.4 | NP_997150.1 | 1 - 2 | 118 | 81 |
GenBank mRNA | AK029707.1 | - | 1 - 2 | 95 | 81 |
BB623661.1 | - | 1 - 2 | 114 | 81 | |
BC096054.1 | - | 1 - 2 | 51 | 81 | |
BE625563.1 | - | 1 - 2 | 43 | 81 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081983.1 | NP_001075452.1 | 1 - 2 | 118 | 81 |